Bovine serum albumin (BSA), Fraction V
Supplier: Roth Carl
Certificates
About this item
- Biotin free
Specifications
- CAS No.:90604-29-8
9048-46-8 - Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
Specifications
Frequently Bought Together