
Indigo (synthetic)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:482-89-3
:
Formula:C₁₆H₁₀N₂O₂
MDL Number:MFCD00005722
:262.27 g/mol
Merck Index:12,04977
Melting Pt:>300 °C
Density:1,48 g/cm³ (20 °C)
EINECS:207-586-9
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Dark blue to violet Powder
- Infrared spectrum:Conforms
- UV spectrum:Complies
Specification Test Results
Frequently Bought Together