Water purification systems, RiOs-DI™ 3
Supplier: Merck
Certificates
About this item
This easy-to-use RiOs-DI™ systems produce Type II pure water whose quality is superior to that of service DI or stills.
- Combination of reverse osmosis (RO) technology and deionisation resins
- Production of up to 15 L purified water per day; integrated 6 liter storage tank
- Small footprint makes it easy to install the RiOs-DI® system wherever you want to in your lab
- User-friendly display provides system status at a glance; the concise Quick Reference Guide is a handy guide for daily operation
- Single SmartPak® RODI cartridge enables easy and rapid replacement
- Available with a built-in 254 nm UV lamp to reduce the level of bacteria for critical applications
Water from RiOs-Di™ systems is suitable for a variety of uses, including buffer and reagent preparation; microbiological culture media preparation; and glassware rinsing.
Ordering information: The purification pack SmartPak™ (171-0752) has to be ordered with the system.
Specifications
- Catalog No:
- 171-1015
- Model:
- RiOs-DI™ 3
- Feed water:
- Potable (tap) water
- Production flow rate:
- 3 L/h @ 15 °C
- TOC content:
- < 30 ppb
- Resistivity at +25 °C:
- > 10 MΩ·cm
- Particles:
- >99 % rejection
- W×D×H:
- 290×330×500 mm
- Weight:
- 7,8 kg