TLC plate heater III
552-0351
: CAMAG
- Heating area:200×200 mm
- Temp.:25...200 °C
- Description:TLC plate heater III, UK-plug
- W×D×H:306×420×34 mm
- Pk:1
The TLC plate heater III is designed for heating TLC plates to a given temperature, while ensuring homogenous heating across the plate. TLC plate heater III has a CERAN® heating surface which is resistant to all common reagents and is easily cleaned. The 200×200 mm heating surface has a grid to facilitate correct positioning of the TLC plate.
- Actual temperature is displayed digitally
- Temperature is selectable between 25 and 200 °C
- Plate heater is protected from overheating