Microcentrifuges, ventilated/refrigerated, 5424 / 5424 R (IVD marked)
Supplier: EPPENDORF
Certificates
About this item
Microcentrifuges with 24 positions, extremely quiet running and perfectly designed for use in all biology and microbiology labs. Available in two versions: with rotary control knobs or touch sensitive keypad.
- Rotary knob version for fast parameter selection
- Keypad version for easy cleaning
- Soft-touch one-finger lid closure for ergonomic operation
- OptiBowl® design for quiet operation, even without a rotor lid
- Centrifuge 5424 R cools down to 4 °C in only 8 minutes
Ordering information: Supplied without rotor or with 24×1,5/2,0 ml standard rotor FA-45-24-11 and fitted lid.
Frequently Bought Together