Universal laboratory centrifuges for sedimentation up to 4×100 ml in swing-out rotors or up to 20000×g in angle rotors.
2-16P:
- Brushless drive, practically maintenance-free
- Stainless steel bowl, illuminated keys for start, stop, lid open
- Speed preselection up to 15000 min⁻¹
- Low speed operation from 100 min⁻¹ possible
- Safety features: Magnetic rotor identification preventing rotors from overspeeding, imbalance switch, no need to open the casing for emergency lid lock release
2-16PK: Refrigerated version with:
- Speed range up to 15300 min⁻¹
- Possibility of precooling the rotors during standstill
- Refrigerant CFC-free
2-16KC: Refrigerated version with:
- Model C with control unit Spincontrol Comfort
- Motorised lid lock
2-16KCH: Heated version of 2-16KC with:
- Temperature control from +40 up to +60 °C depending on rotor and speed
Warranty: one year (centrifuges), two years (accessories).