Cleanroom wipes, MicroSeal® 1200
Supplier: BERKSHIRE
Certificates
About this item
These wipes made from 100% continuous filament polyester knit fabric are designed for the highest level of contamination control in critical processing applications.
- Ideal for cleaning surfaces sensitive to scratching, such as medical devices
- Ultrasonically sealed edge for reduced fiber contamination
- Critically low particles, fibers, ions and extractables
- High abrasion resistance, high absorbency
- Chemically compatible with IPA, Acetone and other solvents
- Weight and caliper provide an excellent hand for cleaning rough surfaces
Ideal for cleaning critical tools, medical devices and machines. Laundered and packed in an ISO 4, Class 10 cleanroom.
Suitable for use in Class 1, ISO 3, M1.5 cleanrooms.
Colour: White
Caution: Data accurate at time of publishing, latest test results and test methods can be found on supplier's website.
Frequently Bought Together