Order Entry
Ireland
ContactUsLinkComponent
Storage vials, Nunc™
Storage vials, Nunc™
Catalog # 391-7589
   Low Manufacturing Impact
undefined
Storage vials, Nunc™
Catalog # 391-7589
   Low Manufacturing Impact

Specifications

  • Cap included:
    Yes
  • Ø×H:
    13×49 mm
  • Capacity:
    2,00 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    NAR Certificate of Retirement
  • Pk:
    500

Specifications

About this item

High purity PPCO (PP copolymer) vial, PE screw cap. Excellent for centrifugation and storage of PCR reagents, restriction enzymes and other diagnostic and biochemical reagents or samples. Available in 0,5 and 2,0 ml sizes with frosted writing area; furthermore 2,0 ml size have graduations at 0,5 and 1,0 ml.

  • Leakproof screw closure even without an O-ring
  • One full turn removes the closure, giving easy access to contents

Can be centrifuged to a max. of 13000×g.

Can be used from −40 to +95 °C.

Can be used from -40 to +95 °C.