Order Entry
Ireland
ContactUsLinkComponent
Tape dispenser
Tape dispenser
Catalog # 819-0003
undefined
Tape dispenser
Catalog # 819-0003

Specifications

  • For Use With:
    Tapes with 25 mm width
  • Colour:
    Black
  • Description:
    Tape dispenser
  • Environmentally Preferable:
  • Pk:
    1

Specifications

About this item

Heavy-duty dispenser suitable for tapes up to 25 mm wide. Accepts 33 and 66 m rolls.

  • Color: Black