Order Entry
Ireland
ContactUsLinkComponent
Marker PAP PEN, Immunostaining
Marker PAP PEN, Immunostaining
Catalog # 720-0169
Supplier:  MUGEN
CAS Number:  
undefined
Marker PAP PEN, Immunostaining
Catalog # 720-0169
Supplier:  MUGEN
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    Marker PAP PEN, Immunostaining
Specifications
About this item
PAP PEN is designed to provide a water repellant barrier when a circle is drawn around a specimen. The barrier of the PAP PEN retains antisera within the defined area and ensures, that only the amount of antibody needed for sufficient reaction is used.
  • PAP PEN stops spreading, reduces waste and divides slides into discret areas for immunostaining, histology, cytology, cell culture, virology, microbiology
  • PAP PEN is effective for immunostaining, PAP method, ABC method, ASD method, enzyme and frozen section methods
  • The staining is unsoluble in acetone and alcohol and can be removed if desired by xylene after the staining procedure