Spoon spatulas
Double ended, broad spoon spatulas made of WIRONIT steel (stainless steel 18/12 with 18% chrome and 10 to 12% nickel).
- Very resistant to corrosion and anti-magnetic
- With one flat spatula end and one spoon end
Frequently Bought Together