Cell culture plates, 96-well, CELLSTAR®
PS, cell culture-treated, sterile. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
- F-bottom wells with excellent optical properties are ideal for microscopic applications
- Clear bottom plates have pigmented walls and a transparent thin film bottom, ideal of fluorescence or luminescence detection methods
- Half area plates allow a reduction of sample volume by up to 50%
- Lid enables gas exchange with minimal evaporation
Colour: µClear® (clear) base, or solid black or white
Colour: µClear® (clear), black or white
- Catalog No:
- 392-0060
- 392-0057
- 392-0056
- 392-0058
- 738-0018
- 392-0022
- 392-0024
- 736-0230
- 738-0019
- 392-0025
- 738-0024
- 392-0023
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
- ISO 14001 (241002); ISO 50001 (241002)
Frequently Bought Together