Safety helmets, uvex airwing B-S-WR
Supplier: UVEX
Certificates
About this item
Ultra-modern design, innovative variable ventilation system, PE safety shell with drainage channel, fabric cap with six fastening points with sliding (or rack) adjustment, side adapter pins (standard width 30 mm) allowing attachment of ear protectors, face shields or Ultravision goggles. Variable ventilation system consisting of adjustable front and rear ventilation screens.
- Variable ventilation system (openings at front and back)
- Rack adjustment
- Short face protection shield for unobstructed field of vision (overhead operations)
Size: 520 - 610 mm; Weight: 360 g
EN 397
Certifications: EN 397
Frequently Bought Together