General purpose gloves, Ultrane 553
Supplier: MAPA
Certificates
About this item
Dark grey polyamide knitted gloves with palm and fingers coated with black nitrile. Gloves feature curved fingers and contoured palm, elastic knitted wrist (natural rubber elastic wrist band) and smooth external surface. Guaranteed silicone free.
- High abrasion resistance for enhanced durability
- Nitrile coating offers superior resistance to oily or greasy components
- Enhanced finger sensitivity with excellent precision of hand movement due to glove design and thickness
- High comfort of seamless knitted liner, palm coated design for improved glove ventilation
These gloves are intended to protect the hand only. They do not protect the wrist or the forearm.
Certifications: EN 388 (4121X), Cat. II
Specifications
- Catalog No:
- 112-0737
- 112-0735
- 112-0738
- 112-0736
- 112-0739
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Finish:
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Coating on palm and fingertips
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388 (4121X)
- EN 388 (4121X)
- EN 388 (4121X)
- EN 388 (4121X)
- EN 388 (4121X)