Chemical resistant gloves, Showa 878
Supplier: Showa
Certificates
About this item
Butyl rubber gloves provide superior resistance to highly corrosive acids and are excellent for handling ketones and esters. Synthetic rubber also provides the highest permeation resistance to gases and water vapours, but does not offer the physical strength of natural rubber.
- Unlined, with smooth finish and rolled cuffs
- Lightweight and flexible
- Impermeable for working in damp or greasy environments
- Cuff prevents dirt from entering the glove
Certifications: Cat III; EN 388:2016+A1:2018; EN ISO 374-1:2016+A1:2018/Type A; EN ISO 374-5:2016.
Specifications
- Catalog No:
- 112-3154
- 112-3266
- 112-3267
- 112-3268
- Model:
- 878
- 878
- 878
- 878
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Smooth
- Smooth
- Smooth
- Smooth
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- EN norm:
- EN 388:2016 (2020X), EN ISO 374-1 Type B (BIK), EN ISO 374-5
- EN 388:2016 (2020X), EN ISO 374-1 Type B (BIK), EN ISO 374-5
- EN 388:2016 (2020X), EN ISO 374-1 Type B (BIK), EN ISO 374-5
- EN 388:2016 (2020X), EN ISO 374-1 Type B (BIK), EN ISO 374-5