Chemical resistant gloves, Ultranitril 492
Supplier: MAPA
Certificates
About this item
These anatomically designed gloves with quality flock lining provide good chemical resistance to hydrocarbon derivatives, alcohols, aromatic and chlorinated solvents.
- Longer working life: Excellent mechanical resistance (abrasion, puncture)
- Double layer process: Enhanced chemical protection
- Recommended for individuals sensitised to natural rubber proteins
Ideal for use in manufacturing industries using cutting oils, metal treatment using solvents, chemical treatments, automotive industries and in the agriculture sector.
Certifications: EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS, ISO 18889 G2, Cat. III
Specifications
- Catalog No:
- 112-6111
- 112-6112
- 112-6115
- 112-6116
- 112-6113
- 112-6114
- Model:
- 492
- 492
- 492
- 492
- 492
- 492
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Embossed
- Embossed
- Embossed
- Embossed
- Embossed
- Embossed
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS
- EN 388 (3101X), EN ISO 374-1 Type A (AJKOPT), EN ISO 374-5 VIRUS