Acid resistant cleanroom gloves, neoprene/nitrile, Advantech 517
Supplier: MAPA
Certificates
About this item
These powder-free cleanroom gloves are made from Tri-polymer blend (nitrile, neoprene, and natural rubber) which offers excellent resistance and strong chemical protection against a great number of chemical products. Free from silicone or paraffin.
- Extreme comfort and durability, exceptional chemical and mechanical resistance
- Resistant to etchants, acids and caustics
- Embossed texture for good grip of wet and dry objects
- Wide cuffs enable double-gloving
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certifications: EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. III; AQL 1,5; ASTM D130.
Packaging: 1 pair/bag; 12 pairs/bag
Specifications
- Catalog No:
- 112-0503
- 112-0500
- 112-0504
- 112-0501
- 112-0502
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- 100/ISO 5
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN 388:2016 (1110X), EN ISO 374-1:2016 Type B (KST), EN ISO 374-5:2016 (MICRO-ORGANISMS)