Industrial gloves, Ultrane 550
Supplier: MAPA
Certificates
About this item
These polyurethane foam gloves with seamless textile support and ventilated back provide light protection.
- Excellent abrasion resistance: longer lasting
- Excellent tactile sensitivity, due to reduced glove thickness
- Absorption of perspiration ensured by the cellular structure of the polyurethane
- Seamless knit lining for additional comfort
Suitable for precision work, such as intricate assembly, sorting small parts, fitting small screws and fasteners, electronic component assembly (displays, LEDs, etc.), high-frequency antennae riveting, microprocessor handling and cosmetics preparation.
Certifications: EN 388:2016 (4131X); Cat. II
Specifications
- Catalog No:
- 112-5625
- 112-5628
- 112-5624
- 112-5626
- 112-5627
- Model:
- 550
- 550
- 550
- 550
- 550
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)
- EN 388:2016 (4131X)