Order Entry
Ireland
ContactUsLinkComponent
VWR® CERTICLEAN® Cleanroom Gloves, Nitrile, Class 10
VWR® CERTICLEAN® Cleanroom Gloves, Nitrile, Class 10
Supplier:  VWR Collection
Certificates
undefined
VWR® CERTICLEAN® Cleanroom Gloves, Nitrile, Class 10
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • VWRU40101-353
  • Ambidextrous / hand-specific:
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Powdered / powder-free:
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4
  • Cuff style:
  • Beaded
  • Finger thickness:
  • 0,16 mm
  • Palm thickness:
  • 0,13 mm