Order Entry
Ireland
ContactUsLinkComponent
Forceps for cover glasses
Forceps for cover glasses
Catalog # 232-1915
Supplier:  BOCHEM
undefined
Forceps for cover glasses
Catalog # 232-1915
Supplier:  BOCHEM
Specifications
  • Type:
    Self-tensioning
  • Length:
    105 mm
  • Pk:
    1
Specifications
About this item
Stainless steel. Self-tensioning, curved ends.
  • For easy, secure handling of cover glasses
  • Can also be used for other laboratory equipment, such as filter discs
  • Easy to clean