Filter holders with receiver, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PS, reusable bottle top vacuum filtration systems. Suitable for analytical or sterile filtration, under vacuum or pressure.
- Graduated receiver
- Two side-arms for tubing 6 to 8 mm inner diameter
- With cover for convenient sterile storage of filtrate
Compatible with 47/50 mm membranes.
Delivery: Supplied with holder and receiver, analytical plate, sterilisation plate and cover.
Specifications
- Catalog No:
- 513-3003
- 513-3002
- 513-3001
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement