Order Entry
Ireland
Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Extraction thimbles
Certificates
Extraction thimbles
Extraction thimbles
13 Options Available Below
Product Details & Documents
Cytiva's Whatman 603G borosilicate glass fiber thimbles with inorganic binder are flexibly designed for use in a full range of commercially available Soxhlet apparatus systems.
  • Resistant to temperatures up to 500 °C (932 °F)
  • Manufactured with walls as thin as 1 to 1,5 mm
  • Analysis of hot and acidic gases, including emission control and smoke stack gas monitoring
  • Soxhlet extraction
  • Analysis of pesticide residues
  • Determination of oil/fat content of foods
  • Analysis of oil and grease in solid wastes
Whatman standard glass extraction thimbles are available in a range of dimensions and wall thicknesses to suit your application.
Product Family Options
Product Information
  • Int.Ø×H
    Thickness
    Availability
    Price
  • 10×38 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 16×50 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 19×90 mm
    1,0 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 22×80 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 23,8×68 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 25×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 28×60 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 240,00 €
    240,00 €
    1 * 25 items
       240,00 €
  • 33×94 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 287,00 €
    287,00 €
    1 * 25 items
       287,00 €
  • 30×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 287,00 €
    287,00 €
    1 * 25 items
       287,00 €
  • 33×100 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 287,00 €
    287,00 €
    1 * 25 items
       287,00 €
  • 33×118 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 364,00 €
    364,00 €
    1 * 25 items
       364,00 €
  • 35×150 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 429,00 €
    429,00 €
    1 * 25 items
       429,00 €
  • 30×80 mm
    1.5 mm
    Log in for availability
    Log in for availability
    1 * 25 items
    1 * 25 items - 209,00 €
    209,00 €
    1 * 25 items
       209,00 €