Order Entry
Ireland
Orders LinkContactUsLinkComponent
Staining boxes with lid
Staining boxes with lid
Staining trough, Clear, Glass, 93×75×55 mm
Catalog # 720-0577
Supplier:   SMITH SCIENTIFIC
Staining boxes with lid
Catalog # 720-0577
Supplier:  SMITH SCIENTIFIC
Staining boxes with lid
Staining boxes with lid
Staining trough, Clear, Glass, 93×75×55 mm
Price
211,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Complete with drop-on lid.
Specifications
  • Colour:
    Clear
  • Description:
    Staining trough
  • Material:
    Glass
  • W×D×H:
    93×75×55 mm