Order Entry
Ireland
ContactUsLinkComponent
VWR® Desiccators, Grease-free
VWR® Desiccators, Grease-free
Catalog # 250-068
Supplier:  JENCONS SCIENTIFIC
VWR® Desiccators, Grease-free
Catalog # 250-068
Supplier:  JENCONS SCIENTIFIC
Specifications
  • Description:
    Spare stopcock, PTFE for vaccum desiccators
  • Seq:
    031
  • Pk:
    1
Specifications
About this item
Borosilicate glass. Elastomer ring is compressed under vacuum making a dry seal. The PTFE valve is situated centrally on the lid, which in conjunction with the dry seal, enables the lid to be lifted on and off vertically.
  • Choice of models with or without Seal-Cote plastic protection, which prevents fragmentation of the glass
  • Lids lift off vertically, giving nearly double the storage volume
Certifications: BS 3423:1962
Delivery: Supplied with sealing ring, gauze, and PTFE stopcock. 9 mm O.D. vacuum connection.