Liebig condenser with standard ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate 3.3 glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours, with fused mantle, cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
DIN 12576
Certifications: DIN 12576.
Frequently Bought Together