Order Entry
Ireland
ContactUsLinkComponent
Hoods, Tyvek® 500, model PH30L0
Hoods, Tyvek® 500, model PH30L0
Catalog # 113-0480
Supplier:  DuPont
undefined
Hoods, Tyvek® 500, model PH30L0
Catalog # 113-0480
Supplier:  DuPont

Specifications

  • Garment application:
    Laboratory
  • Colour:
    White
  • Model:
    PH30L0
  • Pk:
    100

Specifications

About this item

Specially designed for use with Tyvek® apparel, this hood can help offer enhanced protection for body parts that are more exposed to hazardous substances. The hood is made of Tyvek® which is composed of flash spun high density polyethylene. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).

  • Wide flange and elasticated face and neck
  • Ultra-low-linting fabric
  • No silicone added
  • Stitched internal seams
  • Antistatic treatment (EN 1149-5) - on both sides

Typical applications include: pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up, amongst many others.

Certifications: (EU) 2016/425, Cat III, Type PB [6-B]; EN 14126
Packaging: 100 per box, bulk packed.