Order Entry
Ireland
ContactUsLinkComponent
Bottles, wide neck, with screw cap
Bottles, wide neck, with screw cap
  215-0840
undefined
Bottles, wide neck, with screw cap
  215-0840

 

  • Ext.Ø:
    66 mm
  • Thread:
    48/R3
  • Capacity:
    250 ml
  • Height:
    120 mm
  • Pk:
    20

 

 

Brown glass with bakelite closure.

  • Screw closure with paraffin wax lining
  • Good chemical resistance
  • Leakproof

Can be used in all applications involving paste-like or powdery substances or liquids. Protects light sensitive materials.