Burette clamps
Zinc die-cast, nickel plated. For simple, secure fixing of burettes.
- Bosshead for fixing to stand rods
- Jaws spanning distance from 0 to 40 mm
- Clamping jaws with plastic coating to protect burettes
Frequently Bought Together
Zinc die-cast, nickel plated. For simple, secure fixing of burettes.