Order Entry
Ireland
ContactUsLinkComponent
Medical bottles, narrow neck, flat, threaded
Medical bottles, narrow neck, flat, threaded
  HARRWSSF200_G
undefined
Medical bottles, narrow neck, flat, threaded
  HARRWSSF200_G
  • Thread:
    28/R3
  • Capacity:
    200 ml
  • Pk:
    20
Glass, clear, for aluminium screw caps.
  • Narrow neck with short thread
  • Aluminium cap with rubber seal (to be ordered separately)
  • Flat sides