Order Entry
Ireland
Orders LinkContactUsLinkComponent
Gas absorption tubes, Arnold pattern
Gas absorption tubes, Arnold pattern
125 mm
Catalog # 201-0551
Supplier:   JONES FR
Gas absorption tubes, Arnold pattern
Catalog # 201-0551
Supplier:  JONES FR
Gas absorption tubes, Arnold pattern
Gas absorption tubes, Arnold pattern
125 mm
Price
98,80 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With two bulbs.
Specifications
  • Height:
    125 mm