Order Entry
Ireland
ContactUsLinkComponent
Trimethyl orthoacetate, Sigma-Aldrich®
Trimethyl orthoacetate, Sigma-Aldrich®
Supplier:  Merck
Certificates
Trimethyl orthoacetate, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Molar Mass:
    120.14 g/mol
  • Boiling Point:
    108 °C (1013 hPa)
  • Vapor Pressure:
    20 hPa (20 °C)
  • Flash Point:
    15 °C
  • Refractive Index:
    1.3859 (25 °C, 589 nm)
  • Density:
    0.96 g/cm3 (20 °C)

Specification Test Results

Related Information

Frequently Bought Together