Trimethyl orthoacetate 98+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1445-45-0
:
Formula:C₅H₁₂O₃
MDL Number:MFCD00008477
:120.15 g/mol
Boiling Pt:109 °C (1013 hPa)
Melting Pt:–58 °C
Density:0,94 g/cm³
Flash Pt:16 °C
EINECS:215-892-9
UN:3272
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3870 to 1.3900 (20°C, 589 nm)
- GC:≥98.0 %
Specification Test Results
Frequently Bought Together