Order Entry
Ireland
ContactUsLinkComponent
VWR®, Coulometry NewGen Reagent C-FA for coulometric Karl Fischer titration (10x5 ml)
VWR®, Coulometry NewGen Reagent C-FA for coulometric Karl Fischer titration (10x5 ml)
Catalog # 88424.005
Supplier:  VWR Chemicals
New Product
CAS Number:  
undefined
VWR®, Coulometry NewGen Reagent C-FA for coulometric Karl Fischer titration (10x5 ml)
Catalog # 88424.005
Supplier:  VWR Chemicals
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Description:
    NewGen Reagent C-FA for coulometric Karl Fischer titration
  • Seq:
    11
  • Pk Info:
    10x 5 ml

Specifications

About this item

Catholyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, Acetonitrile-based, free of alcohols (free of CMR substances).

  • free of alcohols

  • free of CMR substances
  • Recommended for Li-ion battery electrolytes

Specification:
Suitability (for KF-Titration): complying
Water content: 30 ppm