Stainless Steel Drawers
Supplier: Metro International
New Product
Certificates
About this item
Fully fabricated 304 stainless steel no-flex construction (No pan drop-in required).
- Fully removable drawer with 15,8 kg load capacity per drawer
- Stackable up to 3 high
- NSF listed heavy-duty ball bearing slides
- Drawer face remains flush with worksurface edge regardless of the table depth
Multiple mounting locations: Right, Center, Left (depending on the table length); drawers can be both front facing and rear facing if mounted on the same table.
Frequently Bought Together