Anti-PIN2 Rabbit Polyclonal Antibody (KLH)
Catalog # AGRIAS204521
Supplier: Agrisera
New Product
Specifications
- Antigen name:PIN2
- Clonality:Polyclonal
- Conjugation:KLH (Keyhole Limpet Hemocyanin)
- Host:Rabbit
- Reactivity:A. thaliana
- Format:Lyophilised
- UniProtKB:Q9LU77
- Purity:Immunogen affinity purified serum in PBS pH 7,4
- Molecular weight:69,3 kDa
- Storage temperature:Store lyophilised/reconstituted at −20 °C
- Pk:50 µG
Specifications
About this item
Recommended dilution: 1 : 200 (IL)
Type:
Antigen: PIN2
Clonality: Polyclonal
Clone:
Conjugation: KLH
Epitope:
Host: Rabbit
Isotype:
Reactivity: Arabidopsis thaliana