DURAN® GLS 80® Production and Storage Bottles
Supplier: DWK Life Sciences
New Product
Certificates
About this item
Graduated, manufactured from DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements.
- Thick-walled glass and therefore mechanically more resistant
- The external PU polymer coating provides scratch protection
- The flat, robust base allows the use of large and heavy stir bars
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic
- Easy access to the bottle contents due to the wide diameter of the bottle neck
- Optimum filling and particularly easy cleaning
Ordering information: Supplied as safety coated glass bottle only with a HDPE push-on dust cover. The screw caps and pouring rings must be ordered separately.
Frequently Bought Together