SHOWA XC810 TouchScreen Nitrile, HPPE Gloves
Supplier: Showa
New Product
Certificates
About this item
The new ultra-light, silicone- and fingerprint-free XC810 combines excellent grip, superb tactility, 18-gauge UHMWPE liner providing cut-resistance level D/A4 with touchscreen compatibility for a multitude of industrial applications.
- It has many benefits and features like cut-resistant, flexible, breathable, lightweight
- Increased dexterity and seamless knit
- Form-fitting and abrasion-resistant
- Touch screen compatible
- Natural rubber latex free
- HPPE reinforced and ergonomic
- Microporous nitrile coating
- Embossed grip
It has applications of light assembly of oil-coated pieces, maintenance, mechanical and engineering, repair glass and windows and component assembly. This gloves has cut hazards. This glove is suitable for automotive, construction, glass and mechanical.
Packaging: It contains 12 pairs per polybag/6 polybags per case/72 pairs per case.
Specifications
- Catalog No:
- BESTXC810-8/L
- BESTXC810-7/M
- BESTXC810-10/XXL
- BESTXC810-9/XL
- BESTXC810-6/S
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Gauge:
- G 18
- G 18
- G 18
- G 18
- G 18