Stainless Steel Buckets with Spout
Hygienic, high-quality stainless steel buckets meet international pharma quality compliance regulations. Available in 304 grade stainless steel, these buckets are bright polished throughout, graduated in 1 L intervals, and come complete with a swing handle.
- Buckets available as standard, off-the-shelf for delivery within days
- Buckets in 304 grade stainless steel
- Optional lids available to purchase separately
- Exceptional average surface finish of 0,4 µm Ra
- Can be cleaned with alcohol-based disinfectants, and in industrial dishwashers/autoclaves
- Optional material grade, surface finish, and TSE certification available upon request
A range of optional modifications are available to meet every requirement, including bespoke etching of logos and other information. The complete range includes the following design options: buckets, heavy-duty buckets, and buckets with spout.
Frequently Bought Together