Showa XC510 Cut-Resistant Industrial Gloves
: Showa
New Product
The ultra-light XC510 industrial glove combines an 18-gauge UHMWPE cut-resistant liner with a durable, abrasive-resistant polyurethane palm coating for use in a wide variety of industrial applications.
- Durable, form-fitting, flexible, and lightweight
- Cut-resistance level D/A4
- HPPE reinforced
- Polyurethane palm coating
Flexible and breathable XC510 industrial gloves provide increased dexterity in a form-fitting, seamless knit design. These gloves are recommended for use in maintenance, glass/window repair, component assembly, engineering, and mechanical industries.
: Meets ANSI level A4. EN 388:2016+A1:2018, EN ISO 21420:2020. Cat. II.
: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pairs).
- Catalog No:
- BESTXC510-6/S
- BESTXC510-7/M
- BESTXC510-9/XL
- BESTXC510-8/L
- BESTXC510-10/XXL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Material:
- HPPE
- HPPE
- HPPE
- HPPE
- HPPE
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together