Tyvek® 500 HP coveralls, model TY178S, are specifically designed for workers operating with a full-body harness in environments where hazardous particle or light liquid aerosol hazards are present. Designed to be worn over a full-body harness, the innovative, patent-pending garment design allows workers to access and utilise multiple D-rings on their full-body harness. This garment is compatible with a variety of full-body harness models, making it suitable for a wide range of applications including painting, cleaning, maintenance, and more.
- Three piece hood with elastic face opening
- Stitched seams
- Tyvek® thumb loops
- Tyvek® zipper and Tyvek® storm flap
- Elasticated ankles and wrists and elasticated (glued-in) waist
- Ample crotch area
- Silicone non-added
Innovative, patent-pending design for applications where full-body harness is worn; garment is designed to be worn over the full-body harness. Multiple access points to the full-body harness through the four elasticised D-ring sleeves that are attached to the garment; one in the front, one in the back, and one on each side of the garment. Off-centered zipper entry to accommodate the front D-ring sleeve for front D-ring connection.
Dynamic fall test with an articulated mannequin. Tested in an independent testing laboratory as well as at a harness manufacturer with a conclusion that the Tyvek® 500 HP TY178S HP garment does not affect the dynamic fall test performance of the full-body harness when worn underneath this garment. The test is successful as it demonstrated that the Tyvek® 500 HP TY178S HP garment is not impairing the proper function of the full-body harness in case of a fall.
- Catalog No:
- 114-9766
- 114-9760
- 114-9767
- 114-9762
- 114-9761
- 114-9765
- 114-9763
- 114-9764
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Anti-static:
- +
- +
- +
- +
- +
- +
- +
- +
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Garment application:
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services
- Pharmaceutical, General Purpose, Industrial, Services