Order Entry
Ireland
ContactUsLinkComponent
Silver nitrate 0.01 M (0.01 N) ISO 17034, 17025, 9001
Silver nitrate 0.01 M (0.01 N) ISO 17034, 17025, 9001
Catalog # CMPLZ14941081.1L
New Product
CAS Number:  
Silver nitrate 0.01 M (0.01 N) ISO 17034, 17025, 9001
Catalog # CMPLZ14941081.1L
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :169.87 g/mol
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414