Order Entry
Ireland
ContactUsLinkComponent
Potassium iodide 99.99%, HighPurS-CERT ISO/IEC 17025 certified, ISO 17034
Potassium iodide 99.99%, HighPurS-CERT ISO/IEC 17025 certified, ISO 17034
Catalog # CMPLPSKIC.50G
New Product
CAS Number:  
Potassium iodide 99.99%, HighPurS-CERT ISO/IEC 17025 certified, ISO 17034
Catalog # CMPLPSKIC.50G
New Product
CAS Number:  

Specifications

  • Pk:
    50 g
  • Seq:
    3368
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Dipotassium diiodide, Kali iodide
 :
 :166 g/mol
Storage Temperature:Room Temperature
CAS Number:7681-11-0
Formula:Kl
MDL Number:MFCD00011405
Boiling Pt:1330 °C
Melting Pt:681 °C
Merck Index:13,07727