Order Entry
Ireland
ContactUsLinkComponent
 
Potassium hydroxide 6,6 g/l in ethanol 95% Reag. Ph. Eur. 1070304
Catalog # 87907.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Potassium hydroxide 6,6 g/l in ethanol 95% Reag. Ph. Eur. 1070304
Catalog # 87907.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potash caustic, Caustic potash
 :
 :56.11 g/mol
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553
Boiling Pt:1320
Melting Pt:360
ADR:3 II
Merck Index:12,07806