Sodium chloride 0,9% in aqueous solution
Catalog # DIUKUKF7124
Supplier: DISTRIBUTOR
CAS Number:
Specifications
- Pk:6
- Seq:5055
- Pk Info:6x1 L
Specifications
About this item
:
:58.44 g/mol
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477