Order Entry
Ireland
ContactUsLinkComponent
Pesticide reference standard, β-Endosulfan 100 µg/ml in methanol
Pesticide reference standard, β-Endosulfan 100 µg/ml in methanol
Catalog # ACCUP-092S
Supplier:  AccuStandard
CAS Number:  
Pesticide reference standard, β-Endosulfan 100 µg/ml in methanol
Catalog # ACCUP-092S
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    33213-65-9
  • Description:
    β-Endosulfan 100 µg/ml in methanol
  • Formula:
    C₉H₆Cl₆O₃S
  • Description:
    β-Endosulfan

Specifications

About this item