Pesticide reference standard, α-Endosulfan 100 µg/ml in methanol
Catalog # ACCUP-091S
Supplier: AccuStandard
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:959-98-8
- Description:α-Endosulfan 100 µg/ml in methanol
- Formula:C₉H₆Cl₆O₃S
- Description:β-Endosulfan