Order Entry
Ireland
ContactUsLinkComponent
 
1-Heptanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
 :  Merck
 
undefined
1-Heptanesulphonic acid sodium salt, LiChropur™ for ion pair chromatography, Supelco®
 :  Merck
 

 

  • Hill Formula:
    C₇H₁₅NaO₃S
  • Molar Mass:
    202,24 g/mol
  • Melting Point:
    300 °C
  • pH Value:
    5,5 - 7,5 (100 g/L, H₂O, 20 °C)
  • Bulk Density:
    391 kg/m³
  • Solubility:
    (20 °C) soluble
  • Storage Class:
    10 - 13 Other liquids and solids
  • WGK:
    WGK 1 slightly water endangering

 

Frequently Bought Together