Order Entry
Ireland
ContactUsLinkComponent
2-Thiobarbituric acid
2-Thiobarbituric acid
Catalog # CAYM10009199-1
Supplier:  Cayman Chemical
CAS Number:  
2-Thiobarbituric acid
Catalog # CAYM10009199-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :144.15 g/mol
Boiling Pt:419 °C (1013 hPa)
Melting Pt:235 °C
Density:1,661 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-985-8
CAS Number:504-17-6
Formula:C₄H₄N₂O₂S
MDL Number:MFCD00006674