Order Entry
Ireland
ContactUsLinkComponent
N,N,2,4,6-Pentamethylaniline ≥98%
N,N,2,4,6-Pentamethylaniline ≥98%
Catalog # A10561.14
CAS Number:  
N,N,2,4,6-Pentamethylaniline ≥98%
Catalog # A10561.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

CAS Number:13021-15-3
 :
Formula:C₁₁H₁₇N
MDL Number:MFCD00010596
 :163.26 g/mol
Boiling Pt:213…215 °C
Density:0,907
Flash Pt:78 °C (174 °F)
 :GHS07
Storage Temperature:Ambient