Order Entry
Ireland
Orders LinkContactUsLinkComponent
 

Sudan II

Health
Irritant
Sudan II
Certificates
Sudan II

2 Options Available Below

Product Details & Documents

CAS No.: 3118-97-6 Molecular Formula: C18H16N2O Formula Weight: 276.34 Melting Point: 156-160° MDL No.: MFCD00003896

Synonyms:  1-(2,4-Dimethylphenylazo)-2-naphthol, Solvent Orange 7, 1-((2,4-Dimethylphenyl)diazenyl)naphthalen-2-ol
  • CAS Number:3118-97-6
  • Formula:C₁₈H₁₆N₂O
  • MDL Number:MFCD00003896
  • Molecular Weight:276.34 g/mol
  • Melting Pt:156…160 °C
  • Storage Temperature:Ambient
  • EINECS:221-490-4
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 33,60 €
    33,60 €
    1 * 25 g
       33,60 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 97,00 €
    97,00 €
    1 * 100 g
       97,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.